Learn More
Med Vet International Soda-Lime Prefilled Canister, J0553C, Each
Catalog No. 502522328
Encompass_Preferred
Carbon dioxide absorbent granules. JorVet soda lime is a high-grade granular product that effectively absorbs carbon dioxide that is produced as an anesthesia by-product. It is a mixture of calcium hydroxide Ca(OH)2 also known as lime plus a smaller amount of sodium hydroxide NaOH. Upon exposure to carbon dioxide CO2, the calcium hydroxide converts to calcium carbonate Ca(CO3). This prevents CO2 buildup in a re-breathing circuit. The sodium hydroxide acts as a catalyst in the process. No potassium hydroxide (KOH) is present in JorVet soda lime; this helps to avoid the formation of toxic compounds such as Compound A and carbon monoxide. JorVet soda lime has a pH color change indicator that changes from white to violet when the soda lime needs to be changed. It will eventually return to its original color if not changed. Therefore, a good rule of thumb is to replace the soda lime every time the vaporizer is filled.Soda Lime Cannisters.
Catalog No. 50-252-2328 Supplier Med Vet International Supplier No. JOR553C
Carbon dioxide absorbent granules. JorVet soda lime is a high-grade granular product that effectively absorbs carbon dioxide that is produced as an anesthesia by-product. It is a mixture of calcium hydroxide Ca(OH)2 also known as lime plus a smaller amount of sodium hydroxide NaOH. Upon exposure to carbon dioxide CO2, the calcium hydroxide converts to calcium carbonate Ca(CO3). This prevents CO2 buildup in a re-breathing circuit. The sodium hydroxide acts as a catalyst in the process. No potassium hydroxide (KOH) is present in JorVet soda lime; this helps to avoid the formation of toxic compounds such as Compound A and carbon monoxide. JorVet soda lime has a pH color change indicator that changes from white to violet when the soda lime needs to be changed. It will eventually return to its original color if not changed. Therefore, a good rule of thumb is to replace the soda lime every time the vaporizer is filled.Soda Lime Cannisters.