Learn More
MSC Atrix Canister Vacuum, HEPA Filtration, 6 qt, 1400 W
Catalog No. 502838482
Encompass
This canister vacuum provides powerful 1,400 W cleaning with a three-level HEPA filtration system, making it suitable for home or office use. It features a compact design, a 6 qt HEPA bag, and a 20 ft auto-retractable power cord for easy storage and maneuverability.
  • Three levels of filtration
  • Easy pull HEPA canister
  • Variable speed motor
  • Auto retractable cord
  • Telescopic metal wand with seventeen height settings
  • Indicator light for full HEPA bag
  • 106 CFM
Catalog No. 50-283-8482 Supplier MSC Supplier No. 70912944
This canister vacuum provides powerful 1,400 W cleaning with a three-level HEPA filtration system, making it suitable for home or office use. It features a compact design, a 6 qt HEPA bag, and a 20 ft auto-retractable power cord for easy storage and maneuverability.
  • Three levels of filtration
  • Easy pull HEPA canister
  • Variable speed motor
  • Auto retractable cord
  • Telescopic metal wand with seventeen height settings
  • Indicator light for full HEPA bag
  • 106 CFM