Learn More
Cayman Chemical Cannabibutol, Purity: ≥98%, CAS: 60007-99-0, 5 mg
SDP
Catalog No. 502978834
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; has been found in Cannabis; intended for research and forensic applications
Catalog No. 50-297-8834 Supplier Cayman Chemical Supplier No. 293035 MG
An analytical reference standard that is structurally similar to known phytocannabinoids; has been found in Cannabis; intended for research and forensic applications