Learn More
Sigma Aldrich Fine Chemicals Biosciences Cannabichromenic Acid CBCA
Catalog No. 502901952
Encompass
Cannabichromenic acid (CBCA) is a precursor of CBC biosynthesis a non-psychoactive cannabinoid of Cannabis that reportedly exerts anti-inflammatory antimicrobial and analgesic activity. This Certified Spiking SOLUTIONon(R) is suitable for cannabichromenic acid testing methods by GC/MS LC/MS or HPLC for applications in testing of Cannabis potency or impurity profiling pharmaceutical research and forensic analysis. Cerilliant SOLUTIONon Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient quantitative US DEA-exempt SOLUTIONon format and with TKs for Canadian customers.
Catalog No. 50-290-1952 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C2441ML
Cannabichromenic acid (CBCA) is a precursor of CBC biosynthesis a non-psychoactive cannabinoid of Cannabis that reportedly exerts anti-inflammatory antimicrobial and analgesic activity. This Certified Spiking SOLUTIONon(R) is suitable for cannabichromenic acid testing methods by GC/MS LC/MS or HPLC for applications in testing of Cannabis potency or impurity profiling pharmaceutical research and forensic analysis. Cerilliant SOLUTIONon Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient quantitative US DEA-exempt SOLUTIONon format and with TKs for Canadian customers.