Learn More
Medchemexpress LLC Cannabidiol-C4 | 60113-11-3 | 99.5% | 300.44 | 1 MG
SDP
Catalog No. 5000393549
Encompass_Preferred
Cannabidiol-C4 (CBD-C4) is an impurity in cannabidiol. It is for research use only and not sold to patients.
  • Cytotoxicity against mouse BV-2 cells assessed as reduction in cell viability by crystal violet staining based assay (IC50: 11.1 μM for Compound: 12c)
  • Anti-neuroinflammatory activity in mouse BV-2 cells assessed as inhibition of LPS-induced NO production incubated for 24 hrs by microplate reader analysis (IC50: 5 μM for Compound: 12c)
Catalog No. 50-003-93549 Supplier Medchemexpress LLC Supplier No. HY1694521MG
Cannabidiol-C4 (CBD-C4) is an impurity in cannabidiol. It is for research use only and not sold to patients.
  • Cytotoxicity against mouse BV-2 cells assessed as reduction in cell viability by crystal violet staining based assay (IC50: 11.1 μM for Compound: 12c)
  • Anti-neuroinflammatory activity in mouse BV-2 cells assessed as inhibition of LPS-induced NO production incubated for 24 hrs by microplate reader analysis (IC50: 5 μM for Compound: 12c)