Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ CANNABIS PROCESS CONTROL QC SET
SDP

Catalog No. 23047316 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6 KWIK-STIKS
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-316 6 KWIK-STIKS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-316 Supplier Microbiologics™ Supplier No. 5257P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cannabis Process Control QC Set includes 2 KWIK-STIKs of each strain.

Cannabis Process Control QC Set includes 2 KWIK-STIKs of each strain listed below (6 STIKs total):

  • Aspergillus fumigatus derived from ATCC® 204305™*
  • Escherichia coli (O157:H7) derived from ATCC® 35150™*
  • Salmonella enterica subsp. enterica serovar Typhimurium derived from ATCC® 14028™*

Specifications

Biosafety Level 2
Content And Storage 2°C to 8°C
Format KWIK-STIK
Manufacturing Origin United States
Product Type Quality Control Organism
For Use With (Application) Microbial Identification
Packaging Type Plastic Canister
Quantity 6 KWIK-STIKS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.