Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC AbilityOne Duster, Clear, 10 oz, 2 Pack

Catalog No. 502866089
Encompass

This product is a clear duster designed for use with office equipment.

  • Accessory type: Duster
  • Color: Clear
  • For use with: Office equipment
  • Container size: 10 oz
  • Package amount: 2 pack

Catalog No. 50-286-6089 Supplier MSC Supplier No. 73475162
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a clear duster designed for use with office equipment.

  • Accessory type: Duster
  • Color: Clear
  • For use with: Office equipment
  • Container size: 10 oz
  • Package amount: 2 pack

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.