Learn More
Sigma Aldrich Fine Chemicals Biosciences Cannon-fenske viscometer t1EA
Catalog No. 502901327
Encompass
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of /-0.2% quickly and easily.Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Catalog No. 50-290-1327 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2753011EA
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of /-0.2% quickly and easily.Requires approximately 7 mL of sample. Certificate of Calibration supplied.