Learn More
Canon Instrument Co N2 Viscosity Standard 0.5 L
SDP
Catalog No. NC9898317
Encompass_Preferred
CANNON general purpose standards meet the precision specifications of ASTM D445/D446 and ISO 3104/3105. Lot specific data is provided with each standard.
Catalog No. NC9898317 Supplier Canon Instrument Co Supplier No. 9727C22.016
CANNON general purpose standards meet the precision specifications of ASTM D445/D446 and ISO 3104/3105. Lot specific data is provided with each standard.