Learn More
Cole-Parmer Cannon 9721-R62 Viscometer, Size 1C; 6 to 30 Centistokes
Supplier:  Cole-Parmer 9893414
Encompass_Preferred
Ubbelohde Viscometers. Use to determine kinematic viscosities of transparent Newtonian liquids. Require an 11-mL sample volume. Precision is ±0.2%.
Catalog No. NC1218384