Learn More
Med Vet International Dynarex Nasal Oxygen Cannula Cushion Tip w/Advantage Ear Foam, 14', Adult/Standard Connector, 50/Case
Catalog No. 502831841
Encompass_Preferred
Dynarex Resp-O2 Nasal Oxygen Cannulas with Cushion Tips and Advantage Ear Foam are designed to efficiently deliver supplemental oxygen to respiratory patients. These translucent cannulas come with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility. Adjustable sponge-foam ear wraps and cushioned nasal prongs provide added comfort and help minimize the risk of irritation Includes crush-resistant, star lumen oxygen supply tubing Individually wrapped Not made with natural rubber latex or DEHP
Catalog No. 50-283-1841 Supplier Med Vet International Supplier No. 33175
Dynarex Resp-O2 Nasal Oxygen Cannulas with Cushion Tips and Advantage Ear Foam are designed to efficiently deliver supplemental oxygen to respiratory patients. These translucent cannulas come with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility. Adjustable sponge-foam ear wraps and cushioned nasal prongs provide added comfort and help minimize the risk of irritation Includes crush-resistant, star lumen oxygen supply tubing Individually wrapped Not made with natural rubber latex or DEHP