Learn More
Med Vet International Dynarex Nasal Oxygen Cannula Standard Tip, 14' (4.3 M) Crush Resistant, Adult/Universal Connector, 50/Case
Catalog No. 502831871
Encompass_Preferred
Dynarex Resp-O2 Nasal Oxygen Cannulas with Standard Tips are designed to efficiently deliver supplemental oxygen to respiratory patients. These translucent cannulas are available with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility. Non-flared nasal prongs Crush-resistant, star lumen oxygen supply tubing Individually wrapped Not made with natural rubber latex or DEHP
Catalog No. 50-283-1871 Supplier Med Vet International Supplier No. 33106
Dynarex Resp-O2 Nasal Oxygen Cannulas with Standard Tips are designed to efficiently deliver supplemental oxygen to respiratory patients. These translucent cannulas are available with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility. Non-flared nasal prongs Crush-resistant, star lumen oxygen supply tubing Individually wrapped Not made with natural rubber latex or DEHP