Learn More
Med Vet International Dynarex Etco2 Nasal Sampling Cannula, 7' (2.1 M) Tubing w/Female Luer, infant/Standard 02 Tubing Connector, 25/Case
Catalog No. 502829900
Encompass_Preferred
Dynarex Resp-O2 EtCO2 Nasal Sampling Cannulas with Cushion Nasal Tips are designed to help monitor exhaled carbon dioxide during respiratory treatment. Each soft, cushioned cannula features a permanent barrier between the nasal prongs so that EtCO2 can be sampled from one nostril while oxygen continues to flow to the other nostril. Permanent barrier between nasal prongs allows for EtCO2 sampling with uninterrupted oxygen delivery Constructed from soft materials for added patient comfort Crush-resistant tubing helps prevent occlusion and kinking Features EtCO2 sampling line and O2 tubing of equal length Transparent design allows for clear visualization Not made with natural rubber latex or DEHP
Catalog No. 50-282-9900 Supplier Med Vet International Supplier No. 33183
Dynarex Resp-O2 EtCO2 Nasal Sampling Cannulas with Cushion Nasal Tips are designed to help monitor exhaled carbon dioxide during respiratory treatment. Each soft, cushioned cannula features a permanent barrier between the nasal prongs so that EtCO2 can be sampled from one nostril while oxygen continues to flow to the other nostril. Permanent barrier between nasal prongs allows for EtCO2 sampling with uninterrupted oxygen delivery Constructed from soft materials for added patient comfort Crush-resistant tubing helps prevent occlusion and kinking Features EtCO2 sampling line and O2 tubing of equal length Transparent design allows for clear visualization Not made with natural rubber latex or DEHP