Learn More
World Precision Instrument Cannula and Minipump Holder
SDP
Catalog No. 502539162
Encompass
The 505253 and 505254 models differ in the way they hold the cannula. The 505254 holdes the cannula away from the shaft of the probe holder while the 505253 holds the cannula vertically directly in line with the shaft of the probe holder.
Catalog No. 50-253-9162 Supplier World Precision Instrument Supplier No. 505253
The 505253 and 505254 models differ in the way they hold the cannula. The 505254 holdes the cannula away from the shaft of the probe holder while the 505253 holds the cannula vertically directly in line with the shaft of the probe holder.