Learn More
Sklar Instruments Cannula Instrument Cleaning Brush, Sklar, 5mm x 12 in., Nylon / Stainless Steel. PK/3.
SDP
Catalog No. 501436961 Shop All Sklar Instruments Products
Encompass_Preferred
Cannula Instrument Cleaning Brush, Sklar, 5mm x 12 in., Nylon / Stainless Steel. Sklar's Cannula Instrument Cleaning Brushes are used in the multiple-step process of sterilizing a reusable cannula. Following soaking in enzymatic soluntion the cannula should be cleaned manually with a brush and detergent or ultrasonically. Sklar cannula cleaning brushes are designed for manual cleaning and are available in several sizes in order to properly decontaminate specific size cannulas used for specific procedures. This product has a 5mm wide cleaning brush and an overall length of 12 inches. It also features a loop at the end of the handle for improved user grip and control.
This item is not returnable. View return policy
Catalog No. 50-143-6961 Supplier Sklar Instruments Supplier No. 101354
Cannula Instrument Cleaning Brush, Sklar, 5mm x 12 in., Nylon / Stainless Steel. Sklar's Cannula Instrument Cleaning Brushes are used in the multiple-step process of sterilizing a reusable cannula. Following soaking in enzymatic soluntion the cannula should be cleaned manually with a brush and detergent or ultrasonically. Sklar cannula cleaning brushes are designed for manual cleaning and are available in several sizes in order to properly decontaminate specific size cannulas used for specific procedures. This product has a 5mm wide cleaning brush and an overall length of 12 inches. It also features a loop at the end of the handle for improved user grip and control.