Learn More
Vortex Sales Group 20 Micron Dissolution Cannual Filters (Jar Of 100)
Supplier:  Vortex Sales Group PSFIL20SHR100
Encompass_Preferred
20 micron Dissolution cannula filter, made of UHMW Polyethylene, fits securely on the end of a 1/16” cannula sample probe
Catalog No. NC1881237