Learn More
Quality Lab Accessories Cannula with Kynar Luer Lock, for Dissolution
Catalog No. NC9641290
Encompass
7.75" (195mm) Bent 316 SS 1/8" (3.2mm) OD Cannula with Kynar Luer Lock, Low cost alternative
Catalog No. NC9641290 Supplier Quality Lab Accessories Supplier No. CAN775SHL
7.75" (195mm) Bent 316 SS 1/8" (3.2mm) OD Cannula with Kynar Luer Lock, Low cost alternative