Learn More
Alzet CANNULA HOLDER 1
SDP
Supplier:  Alzet 0008860
Encompass
Designed to hold the removable tab on all ALZET brain infusion cannulae to facilitate stereotaxic placement onto the skull of lab animals. Compatible with the common stereotaxic frames available (i.e., Stoelting, Kopf, WPI, Harvard).
Catalog No. NC0468828