Learn More
RWD LIFE SCIENCE INC CANNULA HOLDER 2.1MM-5.0MM
SDP
Catalog No. NC3391451
Encompass
Compatible with stereotaxic instruments with dual manipulators. Holds electrodes or cannulas of 2.1mm to 5.0mm in diameter.
Catalog No. NC3391451 Supplier RWD LIFE SCIENCE INC Supplier No. 68211
Compatible with stereotaxic instruments with dual manipulators. Holds electrodes or cannulas of 2.1mm to 5.0mm in diameter.