Learn More
Stoelting™ Cannula Holder
SDP
Catalog No. 10000381 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Cannula and Minipump Holder
Cannula holder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-381 Cannula and Minipump Holder
10-000-161 Cannula holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-381 Supplier Stoelting™ Supplier No. 516361
Add to Cart
Add to Cart
Stoelting Cannula Holder was designed to hold 3.4 mm cannula heads, as manufactured by Plastics One™ for cannula sets, and DURECT for ALZET™ Osmotic Pumps
Non-intrusive for cementing cannulas in place.  51636-1 holds the cannula away from the shaft of the probe holder while the 51636 holds the cannula vertically directly in line with the shaft of the probe holder.
Specifications
For Use With (Application) Neuroscience
For Use With (Equipment) Cannulas
Model Cannula and Minipump Holder