Learn More
David Kopf Instruments CANNULA HOLDER
SDP
Catalog No. NC9355237
Encompass
Cannula Holder for use with Alzet/Plastic One cannula 3.4mm/3.5mm dia.
Catalog No. NC9355237 Supplier David Kopf Instruments Supplier No. 1766AP
Cannula Holder for use with Alzet/Plastic One cannula 3.4mm/3.5mm dia.