Learn More
Enterprise Technology Solutions Salter Labs Adult Oral Nasal Sleep
SDP
Catalog No. 502406878
Encompass_Preferred
Each Salter Labs Adult Oral Nasal Sleep Diagnostic Cannula for Simultaneous Monitoring and Sampling (25/Case) features a small, clear 7' (2.13 m) gas sampling tube and a large, green 7' (2.13 m) airflow pressure/snore monitoring tube.Available with a male/female luer lok connector and with or without a hydrophobic/anti-microbial filter, the Adult Oral Nasal Sleep Diagnostic Cannula for Simultaneous Monitoring and Sampling allows for real time pressure/snore monitoring and ETCO2 gas sampling by delivering clear signals and providing accurate results.
Catalog No. 50-240-6878 Supplier Enterprise Technology Solutions Supplier No. SAL50447725
Each Salter Labs Adult Oral Nasal Sleep Diagnostic Cannula for Simultaneous Monitoring and Sampling (25/Case) features a small, clear 7' (2.13 m) gas sampling tube and a large, green 7' (2.13 m) airflow pressure/snore monitoring tube.Available with a male/female luer lok connector and with or without a hydrophobic/anti-microbial filter, the Adult Oral Nasal Sleep Diagnostic Cannula for Simultaneous Monitoring and Sampling allows for real time pressure/snore monitoring and ETCO2 gas sampling by delivering clear signals and providing accurate results.