Learn More
Braintree Scientific Cannula & Mini Pump Holder
SDP
Catalog No. 501953989
Encompass
The Stoelting Cannula Holder was designed to hold 3.4 mm cannula heads, as manufactured by Plastics One™ for cannula sets, and DURECT for ALZET Osmotic Pumps. Non-intrusive for cementing cannulas in place. **Sold for research purposes only and not for human use**
Catalog No. 50-195-3989 Supplier Braintree Scientific Supplier No. 51636
The Stoelting Cannula Holder was designed to hold 3.4 mm cannula heads, as manufactured by Plastics One™ for cannula sets, and DURECT for ALZET Osmotic Pumps. Non-intrusive for cementing cannulas in place. **Sold for research purposes only and not for human use**