Learn More
Instech Mouse tail vein cannula, 29ga stainless steel, 25cm Pebax tube, female luer (ReCathCo 0099EO)
SDP
Catalog No. 50890072
Encompass
The Instech C10SS-MTV1301 is a mouse tail vein cannula featuring a 29ga stainless steel introducer attached to a 25cm (9.8in) Pebax tube for smooth venous access during infusion or sampling procedures. Its bonded female luer provides a secure connection point, while the sharp tip and lightweight tubing support reliable tail vein entry. Supplied with a needle guard and luer cap, it offers low-dead-volume performance suited for precise mouse IV dosing workflows.
Features
  • Mouse tail vein cannula with 29ga stainless steel introducer
  • 25cm (9.8in) Pebax tubing
  • 29ga segment: 0.18x0.33mm dimensions
  • Pebax segment: 0.4x0.8mm dimensions
  • Bonded female luer for secure connection
  • Includes needle guard and luer cap
  • Approx. 35µL dead volume
  • Sharp tip for clean tail vein entry
Catalog No. 50-890-072 Supplier Instech Supplier No. C10SSMTV1301
The Instech C10SS-MTV1301 is a mouse tail vein cannula featuring a 29ga stainless steel introducer attached to a 25cm (9.8in) Pebax tube for smooth venous access during infusion or sampling procedures. Its bonded female luer provides a secure connection point, while the sharp tip and lightweight tubing support reliable tail vein entry. Supplied with a needle guard and luer cap, it offers low-dead-volume performance suited for precise mouse IV dosing workflows.
Features
  • Mouse tail vein cannula with 29ga stainless steel introducer
  • 25cm (9.8in) Pebax tubing
  • 29ga segment: 0.18x0.33mm dimensions
  • Pebax segment: 0.4x0.8mm dimensions
  • Bonded female luer for secure connection
  • Includes needle guard and luer cap
  • Approx. 35µL dead volume
  • Sharp tip for clean tail vein entry