Learn More
Instech Mouse tail vein cannula, 29ga, 25cm Pebax tube, PinPort and injector
SDP
Catalog No. 50890073
Encompass
The Instech C10SS-MTV1417P is a mouse tail vein cannula combining a sharp 29ga stainless steel introducer with a 25cm (9.8in) Pebax extension tube for reliable venous access during sampling or dosing. Its integrated white 25ga PinPort connector enables closed, low-dead-volume handling, and the set includes a PNP3M injector for immediate use. Individually sterilized, it offers smooth insertion performance and dependable connectivity for mouse IV procedures.
Features
  • 29ga stainless steel introducer (0.18x0.33mm)
  • 25cm (9.8in) Pebax tubing (0.4x0.8mm)
  • Integrated white PinPort connector (25ga)
  • Includes PNP3M injector
  • Approx. 30µL dead volume
  • Sharp tip for clean tail vein entry
  • EtO sterilized; individually packaged
Catalog No. 50-890-073 Supplier Instech Supplier No. C10SSMTV1417P
The Instech C10SS-MTV1417P is a mouse tail vein cannula combining a sharp 29ga stainless steel introducer with a 25cm (9.8in) Pebax extension tube for reliable venous access during sampling or dosing. Its integrated white 25ga PinPort connector enables closed, low-dead-volume handling, and the set includes a PNP3M injector for immediate use. Individually sterilized, it offers smooth insertion performance and dependable connectivity for mouse IV procedures.
Features
  • 29ga stainless steel introducer (0.18x0.33mm)
  • 25cm (9.8in) Pebax tubing (0.4x0.8mm)
  • Integrated white PinPort connector (25ga)
  • Includes PNP3M injector
  • Approx. 30µL dead volume
  • Sharp tip for clean tail vein entry
  • EtO sterilized; individually packaged