Learn More
Instech Mouse tail vein cannula, 29ga, 5cm Pebax tube, PinPort and injector
SDP
Catalog No. 50890074
Encompass
The Instech C10SS-MTV1429P is a mouse tail vein cannula combining a sharp 29ga stainless steel introducer with a 5cm (2.0in) Pebax extension tube for quick, controlled venous access. Its integrated 25ga white PinPort connector supports low-dead-volume closed-system dosing, and the set includes a PNP3M injector for immediate PinPort access. Individually EtO sterilized, this cannula offers reliable performance for precise mouse tail vein infusion and sampling workflows.
Features
  • 29ga stainless steel introducer
  • 5cm (2.0in) Pebax tubing (0.4x0.8mm)
  • Integrated 25ga white PinPort connector
  • Includes PNP3M injector
  • Sharp tip for clean tail vein entry
  • Individually packaged and EtO sterilized
Catalog No. 50-890-074 Supplier Instech Supplier No. C10SSMTV1429P
 Restricted Item
Add to Cart
Add to Cart
The Instech C10SS-MTV1429P is a mouse tail vein cannula combining a sharp 29ga stainless steel introducer with a 5cm (2.0in) Pebax extension tube for quick, controlled venous access. Its integrated 25ga white PinPort connector supports low-dead-volume closed-system dosing, and the set includes a PNP3M injector for immediate PinPort access. Individually EtO sterilized, this cannula offers reliable performance for precise mouse tail vein infusion and sampling workflows.
Features
  • 29ga stainless steel introducer
  • 5cm (2.0in) Pebax tubing (0.4x0.8mm)
  • Integrated 25ga white PinPort connector
  • Includes PNP3M injector
  • Sharp tip for clean tail vein entry
  • Individually packaged and EtO sterilized