Learn More
Med Vet International Dynarex Nasal Oxygen Cannula Cushion Tip, 25' (7.6 M) Crush Resistant, Adult/Standard Connector, 25/Case
Catalog No. 502830840
Encompass_Preferred
Dynarex Resp-O2 Nasal Oxygen Cannulas with Cushion Tips are designed to efficiently deliver supplemental oxygen to respiratory patients. The cannulas come with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility. Lightweight cushioned nasal prongs provide added comfort Crush-resistant, star lumen oxygen supply tubing Individually wrapped Not made with natural rubber latex or DEHP
Catalog No. 50-283-0840 Supplier Med Vet International Supplier No. 33128
Dynarex Resp-O2 Nasal Oxygen Cannulas with Cushion Tips are designed to efficiently deliver supplemental oxygen to respiratory patients. The cannulas come with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility. Lightweight cushioned nasal prongs provide added comfort Crush-resistant, star lumen oxygen supply tubing Individually wrapped Not made with natural rubber latex or DEHP