Learn More
Med Vet International Dynarex Nasal Oxygen Cannula Flared Tip, 21' (6.4 M) Crush Resistant, Adult/Standard Connector, 25/Case
Catalog No. 502830833
Encompass_Preferred
Dynarex Resp-O2 Nasal Oxygen Cannulas with Flared Tips are designed to efficiently deliver supplemental oxygen to respiratory patients. The cannulas are available with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility. Flared-tip nasal prongs help ensure a secure fit Crush-resistant, star lumen oxygen supply tubing Individually wrapped Not made with natural rubber latex or DEHP
Catalog No. 50-283-0833 Supplier Med Vet International Supplier No. 33113
Dynarex Resp-O2 Nasal Oxygen Cannulas with Flared Tips are designed to efficiently deliver supplemental oxygen to respiratory patients. The cannulas are available with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility. Flared-tip nasal prongs help ensure a secure fit Crush-resistant, star lumen oxygen supply tubing Individually wrapped Not made with natural rubber latex or DEHP