Learn More
Med Vet International Dynarex Nasal Oxygen Cannula Super Soft-Touch Cushion Tip w/ 14' (4.3M, Adult/Standard Connector), 50/Case
Catalog No. 502829122
Encompass_Preferred
Dynarex Resp-O2 Nasal Oxygen Cannulas, Super Soft Touch with Cushion Tips are designed to efficiently deliver supplemental oxygen to respiratory patients. The cannulas come with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility.
Catalog No. 50-282-9122 Supplier Med Vet International Supplier No. 33145
Dynarex Resp-O2 Nasal Oxygen Cannulas, Super Soft Touch with Cushion Tips are designed to efficiently deliver supplemental oxygen to respiratory patients. The cannulas come with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility.