Learn More
Med Vet International Dynarex Nasal Oxygen Cannula Cushion Tip w/ Ultra Soft Ear Foam w/ 1, Adult/Standard Connector, 50/Case
Catalog No. 502828984
Encompass_Preferred
Dynarex Resp-O2 Nasal Oxygen Cannulas with Cushion Tips and Ultra Soft Ear Foam are designed to efficiently deliver supplemental oxygen to respiratory patients. The cannulas come with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility.
Catalog No. 50-282-8984 Supplier Med Vet International Supplier No. 33155
Dynarex Resp-O2 Nasal Oxygen Cannulas with Cushion Tips and Ultra Soft Ear Foam are designed to efficiently deliver supplemental oxygen to respiratory patients. The cannulas come with either Standard or Universal connectors and feature light green oxygen supply tubing, designed to help improve tube visibility.