Learn More
Med Vet International Dynarex Nasal Oxygen High Flow Cannula w/ 14'(4.3 M) Crush Resistant, Adult/Standard Connector, 25/Case
Catalog No. 502828989
Encompass_Preferred
Dynarex Resp-O2 Nasal Oxygen High Flow Cannulas are designed to deliver supplemental oxygen at high flow rates to patients in respiratory distress. The cannulas come with Standard connectors and feature light green oxygen supply tubing, designed to help improve tube visibility.
Catalog No. 50-282-8989 Supplier Med Vet International Supplier No. 33161
Dynarex Resp-O2 Nasal Oxygen High Flow Cannulas are designed to deliver supplemental oxygen at high flow rates to patients in respiratory distress. The cannulas come with Standard connectors and feature light green oxygen supply tubing, designed to help improve tube visibility.