Learn More
Instech Rat tail vein cannula, 25ga, 6cm PE/PVC tube, PinPort and injector
SDP
Catalog No. 50890076
Encompass
The Instech C15SS-RTV1453P is a rat tail vein cannula featuring a sharp 25ga stainless steel introducer paired with a 6cm (2.4in) PE/PVC extension tube for controlled venous access during dosing or sampling. Its integrated white 25ga PinPort connector enables a closed, low-dead-volume workflow, and each set includes a PNP3M injector and needle guard for safe, ready-to-use operation. Individually EtO sterilized, it provides dependable performance for precise rat tail vein procedures.
Features
  • 25ga stainless steel introducer (0.25x0.51mm)
  • 6cm (2.4in) PE/PVC tubing (0.4x1.3mm)
  • Integrated 25ga white PinPort connector
  • Includes PNP3M injector and needle guard
  • Sharp tip for clean tail vein entry
  • Individually packaged and EtO sterilized
Catalog No. 50-890-076 Supplier Instech Supplier No. C15SSRTV1453P
The Instech C15SS-RTV1453P is a rat tail vein cannula featuring a sharp 25ga stainless steel introducer paired with a 6cm (2.4in) PE/PVC extension tube for controlled venous access during dosing or sampling. Its integrated white 25ga PinPort connector enables a closed, low-dead-volume workflow, and each set includes a PNP3M injector and needle guard for safe, ready-to-use operation. Individually EtO sterilized, it provides dependable performance for precise rat tail vein procedures.
Features
  • 25ga stainless steel introducer (0.25x0.51mm)
  • 6cm (2.4in) PE/PVC tubing (0.4x1.3mm)
  • Integrated 25ga white PinPort connector
  • Includes PNP3M injector and needle guard
  • Sharp tip for clean tail vein entry
  • Individually packaged and EtO sterilized