Learn More
Med Vet International Cannulated Drill, 2.7mm x 130mm, w/1.25mm Cannulation, J0372CNB
Catalog No. 5000584783
Encompass_Preferred
Cannulated Drills. For Maximum control over the alignment of bone tunnels use of cannulated drills is preferred. A fine guide wire is drilled through where the bone tunnel or screw hole is required. Only when the surgeon is satisfied that the guide wire i
Catalog No. 50-005-84783 Supplier Med Vet International Supplier No. JOR372CNB
Cannulated Drills. For Maximum control over the alignment of bone tunnels use of cannulated drills is preferred. A fine guide wire is drilled through where the bone tunnel or screw hole is required. Only when the surgeon is satisfied that the guide wire i