Learn More
Med Vet International Cannulated Drill, 3.5mm x 130mm, w/1.5mm Cannulation, J0372CNC
Catalog No. 5000584442
Encompass_Preferred
Cannulated Drills. For maximum control over the alignment of bone tunnels use of cannulated drills is preferred. A fine gui de wire is drilled through where the bone tunnel or screw hole is required. Only when the surgeon is satisfied that the guide wire
Catalog No. 50-005-84442 Supplier Med Vet International Supplier No. JOR372CNC
Cannulated Drills. For maximum control over the alignment of bone tunnels use of cannulated drills is preferred. A fine gui de wire is drilled through where the bone tunnel or screw hole is required. Only when the surgeon is satisfied that the guide wire