Learn More
Braintree Scientific Cannulock Catheter Access Button Caps, Green Magnetic
SDP
Catalog No. 501954503
Encompass
Cannulock™ Catheter Access Buttons™ and Ports Quick, clean catheter access for secure, non-slip catheter connections Catheter Access Button™ Caps Lightweight metal caps connect and protect Catheter Access Buttons™, allowing users to group house animals without worry Magnetic cap is self-aligning, attaching easily Lightweight and durable for worry free group housing Three colors available for easy identification of study groups **Sold for research purposes only and not for human use**
Catalog No. 50-195-4503 Supplier Braintree Scientific Supplier No. CABGCR
Cannulock™ Catheter Access Buttons™ and Ports Quick, clean catheter access for secure, non-slip catheter connections Catheter Access Button™ Caps Lightweight metal caps connect and protect Catheter Access Buttons™, allowing users to group house animals without worry Magnetic cap is self-aligning, attaching easily Lightweight and durable for worry free group housing Three colors available for easy identification of study groups **Sold for research purposes only and not for human use**