Learn More
Braintree Scientific Cannulock Catheter Access Port, 22g, blue, 100/pk
SDP
Catalog No. 501954519
Encompass
Cannulock™ Catheter Access Buttons™ and Ports Quick, clean catheter access for secure, non-slip catheter connections Catheter Access Ports™ Safe and secure rat Catheter Access Buttons™ - smooth barbed connector locks with a catheter to eliminate the risk of slipping or disconnecting Soft Dacron felt for secure anchoring and rapid tissue ingrowth Each sterile pouch contains 2 ports Quick, clean access to 1, 2, or 3 catheters in rats, hamsters and guinea pigs 22G catheter connection fits multiple vascular access catheters: jugular, femoral, carotid, gastric, and bile Closed, split septum access to minimize introduction of pathogens Magnetic tether attaches quickly and easily for a simplified connection Lightweight metal cap protects the button for worry free group housing Built to withstand multiple insertions **Sold for research purposes only and not for human use**
Catalog No. 50-195-4519 Supplier Braintree Scientific Supplier No. CLP22B100
Cannulock™ Catheter Access Buttons™ and Ports Quick, clean catheter access for secure, non-slip catheter connections Catheter Access Ports™ Safe and secure rat Catheter Access Buttons™ - smooth barbed connector locks with a catheter to eliminate the risk of slipping or disconnecting Soft Dacron felt for secure anchoring and rapid tissue ingrowth Each sterile pouch contains 2 ports Quick, clean access to 1, 2, or 3 catheters in rats, hamsters and guinea pigs 22G catheter connection fits multiple vascular access catheters: jugular, femoral, carotid, gastric, and bile Closed, split septum access to minimize introduction of pathogens Magnetic tether attaches quickly and easily for a simplified connection Lightweight metal cap protects the button for worry free group housing Built to withstand multiple insertions **Sold for research purposes only and not for human use**