Learn More
Braintree Scientific Cannulock Catheter Access Port Adapter, 20/pk
SDP
Catalog No. 501954551
Encompass
Cannulock™ Catheter Access Buttons™ and Ports Quick, clean catheter access for secure, non-slip catheter connections Catheter Access Port Adapters™ Provides a remarkably secure connection between catheter and the port Each sterile pouch contains 2 adapters Connect to a slip tip syringe for easy catheter access Available pre-attached to empty syringe or on pre-filled flush and lock syringes, and in customized kits for maximum efficiency (email requirements for quote) **Sold for research purposes only and not for human use**
Catalog No. 50-195-4551 Supplier Braintree Scientific Supplier No. CLPAD20
 Restricted Item
Add to Cart
Add to Cart
Cannulock™ Catheter Access Buttons™ and Ports Quick, clean catheter access for secure, non-slip catheter connections Catheter Access Port Adapters™ Provides a remarkably secure connection between catheter and the port Each sterile pouch contains 2 adapters Connect to a slip tip syringe for easy catheter access Available pre-attached to empty syringe or on pre-filled flush and lock syringes, and in customized kits for maximum efficiency (email requirements for quote) **Sold for research purposes only and not for human use**