Learn More
Enterprise Technology Solutions Canon 8-Dgt Fold Pocket Calculator,Dual Pwr,4-1/3"x2-2/3"x3/5",CCL
SDP
Catalog No. 5000739831
Encompass_Preferred
Foldout wallet calculator features an eight-digit, liquid crystal display (LCD) and four-key memory. Mathematical features include square root, percentage, sign change, and variable decimal. Fold-over design and sturdy hard cover casing offer protection and convenience during transport. 2 oz. calculator runs on battery and solar power and automatically shuts down after seven minutes of nonuse.
Catalog No. 50-007-39831 Supplier Enterprise Technology Solutions Supplier No. CNMLS555H
Foldout wallet calculator features an eight-digit, liquid crystal display (LCD) and four-key memory. Mathematical features include square root, percentage, sign change, and variable decimal. Fold-over design and sturdy hard cover casing offer protection and convenience during transport. 2 oz. calculator runs on battery and solar power and automatically shuts down after seven minutes of nonuse.