Learn More
Enterprise Technology Solutions Canon Photo Paper,ZINK,Glossy,f/IVY Printers,2"x3",50 Sheets,White
SDP
Catalog No. 5000830787
Encompass_Preferred
ZINK Photo Paper features "ZERO INK" printing technology. Instead of ink, it uses colorful, dye-based crystals embedded inside the paper. Before printing, the embedded dye crystals are clear so ZINK Paper looks like regular, white photo paper. Your ZINK-enabled device uses heat to activate and colorize these crystals. Glossy paper has a peel-and-stick back for easy placement anywhere. Printed paper is smudge-free, water-resistant and tear-resistant. ZINK Photo Paper is compatible with Canon IVY CLIQ2, IVY CLIQ+2 and IVY Mini Photo Printers that come in multiple fun colors.
Catalog No. 50-008-30787 Supplier Enterprise Technology Solutions Supplier No. CNMZINCPAPER50
ZINK Photo Paper features "ZERO INK" printing technology. Instead of ink, it uses colorful, dye-based crystals embedded inside the paper. Before printing, the embedded dye crystals are clear so ZINK Paper looks like regular, white photo paper. Your ZINK-enabled device uses heat to activate and colorize these crystals. Glossy paper has a peel-and-stick back for easy placement anywhere. Printed paper is smudge-free, water-resistant and tear-resistant. ZINK Photo Paper is compatible with Canon IVY CLIQ2, IVY CLIQ+2 and IVY Mini Photo Printers that come in multiple fun colors.