Learn More
Medchemexpress LLC Canrenoate potassium | 2181-04-6 | 99.9% | 100 MG
SDP
Catalog No. 5000412763
Encompass_Preferred
Canrenoate potassium is a proagent that releases canrenone. It acts as a potent, competitive mineralocorticoid receptor (aldosterone receptor) antagonist. As a diuretic, it is used for hypertension research.
  • Proagent that releases canrenone
  • Potent, competitive mineralocorticoid receptor antagonist
  • Used in hypertension research as a diuretic
Catalog No. 50-004-12763 Supplier Medchemexpress LLC Supplier No. HYB1582A100MG
Canrenoate potassium is a proagent that releases canrenone. It acts as a potent, competitive mineralocorticoid receptor (aldosterone receptor) antagonist. As a diuretic, it is used for hypertension research.
  • Proagent that releases canrenone
  • Potent, competitive mineralocorticoid receptor antagonist
  • Used in hypertension research as a diuretic