Learn More
Medchemexpress LLC Canrenoate potassium (aldadiene potassium) | 2181-04-6 | 99.9% | 500 MG
SDP
Catalog No. 5000413206
Encompass_Preferred
Canrenoate (Aldadiene) potassium is a proagent that releases canrenone. It is a potent, competitive mineralocorticoid receptor (aldosterone receptor) antagonist. Potassium canrenoate, as a diuretic, is used for the research of hypertension.
  • Proagent that releases canrenone
  • Potent, competitive mineralocorticoid receptor antagonist
  • Used for the research of hypertension
Catalog No. 50-004-13206 Supplier Medchemexpress LLC Supplier No. HYB1582A500MG
Canrenoate (Aldadiene) potassium is a proagent that releases canrenone. It is a potent, competitive mineralocorticoid receptor (aldosterone receptor) antagonist. Potassium canrenoate, as a diuretic, is used for the research of hypertension.
  • Proagent that releases canrenone
  • Potent, competitive mineralocorticoid receptor antagonist
  • Used for the research of hypertension