Learn More
Medchemexpress LLC Canrenone (Aldadiene) | 976-71-6 | 99.6% | 100 MG
SDP
Catalog No. 5000412708
Encompass_Preferred
Canrenone (Aldadiene) is an aldosterone antagonist extensively used as a diuretic agent. It inhibits the production of eortieosterone, 18-hydroxydesoxyeortieosterone, 18-hydroxycorticosterone, and aldosterone in a dose-dependent manner. Canrenone also reduces platelet-derived growth factor-induced cell proliferation and motility, and inhibits the activity of the Na+/H+ exchanger 1 induced by platelet-derived growth factor. It is the principal active metabolite of Spironolactone.
  • Aldosterone antagonist
  • Endogenous metabolite
  • Solid appearance
  • White to light yellow color
  • Steroid structure classification
  • Used as a diuretic agent
  • Inhibits the production of several corticosteroids
  • Reduces cell proliferation and motility induced by platelet-derived growth factor
  • Inhibits Na+/H+ exchanger 1 activity
Catalog No. 50-004-12708 Supplier Medchemexpress LLC Supplier No. HYB1438100MG
Canrenone (Aldadiene) is an aldosterone antagonist extensively used as a diuretic agent. It inhibits the production of eortieosterone, 18-hydroxydesoxyeortieosterone, 18-hydroxycorticosterone, and aldosterone in a dose-dependent manner. Canrenone also reduces platelet-derived growth factor-induced cell proliferation and motility, and inhibits the activity of the Na+/H+ exchanger 1 induced by platelet-derived growth factor. It is the principal active metabolite of Spironolactone.
  • Aldosterone antagonist
  • Endogenous metabolite
  • Solid appearance
  • White to light yellow color
  • Steroid structure classification
  • Used as a diuretic agent
  • Inhibits the production of several corticosteroids
  • Reduces cell proliferation and motility induced by platelet-derived growth factor
  • Inhibits Na+/H+ exchanger 1 activity