Learn More
Medchemexpress LLC Canrenone-d6 (Aldadiene-d6) | 976-71-6 | 98.8% | 1 MG
SDP
Catalog No. 5000413393
Encompass_Preferred
Canrenone-d6 is the deuterium-labeled version of Canrenone. Canrenone (Aldadiene) is an aldosterone antagonist frequently used as a diuretic agent. It is intended for research use only.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13393 Supplier Medchemexpress LLC Supplier No. HYB1438S1MG
Canrenone-d6 is the deuterium-labeled version of Canrenone. Canrenone (Aldadiene) is an aldosterone antagonist frequently used as a diuretic agent. It is intended for research use only.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs.