Learn More
Medchemexpress LLC Canrenone-d7 (aldadiene-d7) | 976-71-6 | MFCD00071735 | 99.6% | 347.50 | C22H21D7O3 | 10 MG
SDP
Catalog No. 5000297267
Encompass_Preferred
Canrenone-d7 is a deuterium-labeled analogue of canrenone, an aldosterone antagonist commonly used as a diuretic. This isotopically labeled standard is supplied at high purity for analytical and research applications and is intended for use as a tracer or internal standard in quantitative analyses by NMR, GC-MS, or LC-MS.
  • High isotopic labeling and purity for accurate quantitation.
  • Suitable as a tracer or internal standard in NMR, GC-MS, and LC-MS.
  • Supplied in small research-scale quantities for analytical workflows.
  • Intended for research use only; not for human or therapeutic use.
  • Stable when stored and handled according to safety data sheet recommendations.
Catalog No. 50-002-97267 Supplier Medchemexpress LLC Supplier No. HYB1438S210MG
Canrenone-d7 is a deuterium-labeled analogue of canrenone, an aldosterone antagonist commonly used as a diuretic. This isotopically labeled standard is supplied at high purity for analytical and research applications and is intended for use as a tracer or internal standard in quantitative analyses by NMR, GC-MS, or LC-MS.
  • High isotopic labeling and purity for accurate quantitation.
  • Suitable as a tracer or internal standard in NMR, GC-MS, and LC-MS.
  • Supplied in small research-scale quantities for analytical workflows.
  • Intended for research use only; not for human or therapeutic use.
  • Stable when stored and handled according to safety data sheet recommendations.