Learn More
Medchemexpress LLC CANT1 Human HEK293 1mg
SDP
Catalog No. 5000178779
Encompass_Preferred
CANT1 is a calcium-dependent nucleotidase that shows a clear preference for UDP and the order of its enzymatic activity is UDP GDP UTP GTP It is worth noting that it has little activity on ADP and even less activity on ATP but it does not hydrolyze AMP and GMP CANT1 Protein Human (HEK293 His) is the recombinant human-derived CANT1 protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-78779 Supplier Medchemexpress LLC Supplier No. HYP743551MG
Add to Cart
Add to Cart
CANT1 is a calcium-dependent nucleotidase that shows a clear preference for UDP and the order of its enzymatic activity is UDP GDP UTP GTP It is worth noting that it has little activity on ADP and even less activity on ATP but it does not hydrolyze AMP and GMP CANT1 Protein Human (HEK293 His) is the recombinant human-derived CANT1 protein expressed by HEK293 with N-His labeled tag