Learn More
Medchemexpress LLC CANT1 Human HEK293 50ug
SDP
Catalog No. 5000178780
Encompass_Preferred
CANT1 is a calcium-dependent nucleotidase that shows a clear preference for UDP and the order of its enzymatic activity is UDP GDP UTP GTP It is worth noting that it has little activity on ADP and even less activity on ATP but it does not hydrolyze AMP and GMP CANT1 Protein Human (HEK293 His) is the recombinant human-derived CANT1 protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-78780 Supplier Medchemexpress LLC Supplier No. HYP7435550UG
CANT1 is a calcium-dependent nucleotidase that shows a clear preference for UDP and the order of its enzymatic activity is UDP GDP UTP GTP It is worth noting that it has little activity on ADP and even less activity on ATP but it does not hydrolyze AMP and GMP CANT1 Protein Human (HEK293 His) is the recombinant human-derived CANT1 protein expressed by HEK293 with N-His labeled tag