Learn More
Enzo Life Sciences Cantharidic acid (50mg). CAS: 28874-45-5
SDP
Supplier:  Enzo Life Sciences BMLPR1060050
Encompass_Preferred
Potent and selective inhibitor of PP2A, IC50=53nM (IC50 for PP1=562nM). Useful for inhibiting PP1 and PP2A during purification of phosphorylated proteins. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or ethanol (10mg/ml warm). Long Term Storage: Ambient.
Catalog No. 50-201-1504