Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cantharidic acid disodium | 1465-77-6 | 95.0% | C10H12Na2O5 | 10MG
SDP

Catalog No. 5000259018
Encompass_Preferred

Cantharidic acid disodium is the disodium salt of cantharidic acid and a biochemical reagent that selectively inhibits protein phosphatases PP2A and PP1. It is used in research to probe phosphatase-regulated signaling and apoptosis pathways and is supplied as a solid for laboratory assays.

  • Selective inhibitor of protein phosphatases PP2A and PP1.
  • Induces apoptosis in various human cell lines, useful for signaling studies.
  • Supplied as a solid suitable for biochemical and cell-based assays.
  • Molecular formula C10H12Na2O5 and molecular weight 258.18 g/mol.
  • Purity reported as 95.0%.
  • Available in small research pack sizes such as 5 mg and 10 mg.

Catalog No. 50-002-59018 Supplier Medchemexpress LLC Supplier No. HY137135A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cantharidic acid disodium is the disodium salt of cantharidic acid and a biochemical reagent that selectively inhibits protein phosphatases PP2A and PP1. It is used in research to probe phosphatase-regulated signaling and apoptosis pathways and is supplied as a solid for laboratory assays.

  • Selective inhibitor of protein phosphatases PP2A and PP1.
  • Induces apoptosis in various human cell lines, useful for signaling studies.
  • Supplied as a solid suitable for biochemical and cell-based assays.
  • Molecular formula C10H12Na2O5 and molecular weight 258.18 g/mol.
  • Purity reported as 95.0%.
  • Available in small research pack sizes such as 5 mg and 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.