Learn More
Apexbio Technology LLC Cantharidin 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092595
Encompass_Preferred
Cantharidin is a naturally occurring terpenoid compound known to function as a potent and selective inhibitor of serine/threonine protein phosphatases particularly protein phosphatase 2A (PP2A) and protein phosphatase 1 (PP1) By inhibiting these enzymes cantharidin disrupts the regulation of numerous cellular processes including cell cycle progression apoptosis and signal transduction pathways This molecule has been widely utilized in research to investigate phosphatase-mediated signaling mechanisms and to explore its potential role in modulating cancer cell proliferation and death Cantharidin serves as a valuable tool for dissecting the functional roles of PP1 and PP2A in various biological systems
Catalog No. 50-000-92595 Supplier Apexbio Technology LLC Supplier No. N16865.1
Cantharidin is a naturally occurring terpenoid compound known to function as a potent and selective inhibitor of serine/threonine protein phosphatases particularly protein phosphatase 2A (PP2A) and protein phosphatase 1 (PP1) By inhibiting these enzymes cantharidin disrupts the regulation of numerous cellular processes including cell cycle progression apoptosis and signal transduction pathways This molecule has been widely utilized in research to investigate phosphatase-mediated signaling mechanisms and to explore its potential role in modulating cancer cell proliferation and death Cantharidin serves as a valuable tool for dissecting the functional roles of PP1 and PP2A in various biological systems