Learn More
AdipoGen Cantharidin
SDP
Catalog No. 502561355
Encompass_Preferred
Chemical. CAS 56-25-7. Formula C10H12O4. MW 196.2. . Cantharidin is a natural toxin produced by blister beetles. It is a selective protein phosphatase 2A PP2A inhibitor and weakly inhibits protein phosphatases 1 PP1. Displays > 500-fold selectivity over PP2B. Induces G2/M phase arrest and apoptosis, suppresses autophagy and inhibits cell proliferation. It is used topically against Molluscum contagiosum virus MCV infections and associated warts. Has shown potent anticancer activities on many types of human cancer cells.
Catalog No. 50-256-1355 Supplier AdipoGen Supplier No. CDXC0643M500
Chemical. CAS 56-25-7. Formula C10H12O4. MW 196.2. . Cantharidin is a natural toxin produced by blister beetles. It is a selective protein phosphatase 2A PP2A inhibitor and weakly inhibits protein phosphatases 1 PP1. Displays > 500-fold selectivity over PP2B. Induces G2/M phase arrest and apoptosis, suppresses autophagy and inhibits cell proliferation. It is used topically against Molluscum contagiosum virus MCV infections and associated warts. Has shown potent anticancer activities on many types of human cancer cells.